missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Reagent Grade, ≥99%, Honeywell

Reagent Grade,≥99%

32.90 € - 510.28 €

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O
View More Specs

Products 6
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15627800
View Documents Promotion Details
Honeywell Research Chemicals
D158550-500ML
500 mL Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15677790
View Documents Promotion Details
Honeywell Research Chemicals
D158550-1L
1 L Poly Bottle
88.74 €
1 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15687790
View Documents Promotion Details
Honeywell Research Chemicals
D158550-2.5L
2.5 L Poly Bottle
153.27 €
2.50 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15637800
View Documents Promotion Details
Honeywell Research Chemicals
D158550-5L
5 L Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15647800
View Documents Promotion Details
Honeywell Research Chemicals
D158550-6X1L
6 x 1 L Poly Bottle
462.91 €
Pack of 6
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15617800
View Documents Promotion Details
Honeywell Research Chemicals
D158550-4X2.5L
4 x 2.5 L Poly Bottle
510.28 €
Pack of 4
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

68-12-2
73.10
ZMXDDKWLCZADIW-UHFFFAOYSA-N
CHEBI:17741
CN(C)C=O
C3H7NO
MFCD00003284
6228
N,N-dimethylformamide

Specifications

-61°C (literature)
≥90.0000%
C3H7NO
MFCD00003284
605365
ZMXDDKWLCZADIW-UHFFFAOYSA-N
N,N-dimethylformamide
6228
73.10
Reagent
152°C to 153°C
57.5°C
2.7 mmHg (20°C)
N,N-Dimethylformamide, Reagent Grade, ≥99%
68-12-2
≤100.0000%
HCON(CH3)2
UN2265
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
CN(C)C=O
73.10
CHEBI:17741
>99%
0.944 g/mL(lit.)
0.152
833°F
max. 0.1%
Videos
Safety and Handling

Safety and Handling

P201-P210-P261-P280-P308 + P313-P370 + P378

missing translation for 'shelfLife' : 1080 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: N,N-Dimethylformamide

missing translation for 'einecsNumber' : 200-679-5

missing translation for 'rtecsNumber' : LQ2100000

Recommended Storage : Room Temp

Product Identifier
  • N,N-Dimethylformamide

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications