missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals N,N-Dimethylglycine hydrochloride, 99% Product Code.: 10431441

Thermo Scientific Chemicals N,N-Dimethylglycine hydrochloride, 99%

Product Code. 10431441
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10431441

Brand: Thermo Scientific Chemicals 204660050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-06-7
Molecular Formula C4H9NO2·HCl
Molecular Weight (g/mol) 139.58
MDL Number MFCD00012610
InChI Key FKASAVXZZLJTNX-UHFFFAOYSA-N
Synonym n,n-dimethylglycine hydrochloride, dimethylglycine hydrochloride, glycine, n,n-dimethyl-, hydrochloride, n-methylsarcosine hydrochloride, n,n-dimethylglycine hcl, dimethylamino acetic acid hydrochloride, unii-yxk75eae92, yxk75eae92, n,n-dimethylaminoacetic acid hydrochloride, 2-dimethylamino acetic acid hydrochloride
PubChem CID 75605
IUPAC Name 2-(dimethylamino)acetic acid;hydrochloride
SMILES CN(C)CC(=O)O.Cl

Specifications

Melting Point 188°C to 191°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CO2H·HCl
Beilstein 04, I, 469
Merck Index 15, 3269
Solubility Information Solubility in water: 100g/L (20 c). Other solubilities: insoluble in chloroform and acetone
Formula Weight 139.58
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material N, N-Dimethylglycine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453368 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number204661000,204660250,204660050
Lot NumberA0396523 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number204661000,204660250,204660050
Lot NumberA0435219 Certificate TypeCertificate of Analysis Date30/10/2021 Catalog Number204661000,204660250,204660050
Lot NumberA0403617 Certificate TypeCertificate of Analysis Date21/06/2021 Catalog Number204661000,204660250,204660050
Product Identifier
  • N,N-Dimethylglycine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N,N-Dimethylglycine hydrochloride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.