missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyloctadecylamine, 89%, tech., Thermo Scientific Chemicals

48.04 € - 148.74 €

Chemical Identifiers

CAS 124-28-7
Molecular Formula C20H44ClN
Molecular Weight (g/mol) 334.03
MDL Number MFCD00048496
InChI Key PFKRTWCFCOUBHS-UHFFFAOYSA-N
Synonym n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PubChem CID 15365
IUPAC Name N,N-dimethyloctadecan-1-amine
SMILES [Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10645681
View Documents
Thermo Scientific Acros
408432500
250 g Glass bottle
48.04 €
250g
Please to purchase this item. Need a web account? Register with us today!
10549472
View Documents
Thermo Scientific Acros
408430010
1 kg Glass bottle
148.74 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-28-7
334.03
PFKRTWCFCOUBHS-UHFFFAOYSA-N
15365
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
C20H44ClN
MFCD00048496
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
N,N-dimethyloctadecan-1-amine

Specifications

124-28-7
100.0
Colorless to Yellow
347.0°C
Authentic
C20H44ClN
MFCD00048496
0.8
Solubility in water: slightly soluble
[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
334.03
297.57
Technical
N, N-Dimethyloctadecylamine, tech., 89%
84.0
15.0°C to 20.0°C
0.8000g/mL
155°C
84% min. (C18) (GC)
(CH3)2N(CH2)17CH3
15, 3522
n,n-dimethyloctadecylamine, dymanthine, dimantine, n,n-dimethyl-n-octadecylamine, dimethyloctadecylamine, 1-octadecanamine, n,n-dimethyl, n,n-dimethyl-1-octadecanamine, dimethyl stearamine, stearyldimethylamine, n,n-dimethylstearylamine
PFKRTWCFCOUBHS-UHFFFAOYSA-N
N,N-dimethyloctadecan-1-amine
15365
89%
Liquid After Melting
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-694-8

missing translation for 'rtecsNumber' : RG4200000

missing translation for 'tsca' : TSCA

Product Identifier
  • N,N-Dimethyloctadecylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only