missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, ≥99%, for biochemistry, for electrophoresis

Product Code. 10226120
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym MBA
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Absorbance (1% aq. soln.) (1 cm cell), (1% aq. soln.) (1 cm cell) at 0nm, 0.2 max. at 290nm
Physical Form Fine Needle-like Crystals or Crystalline Powder
Acrylic Acid 0.02% max.
CAS Min % 99.0
CAS Max % 100.0
Density 5.31 (Air = 1.0)
Color White
pH 5 (2.5% min. aq. soln.)
Quantity 25 g
Assay Percent Range 99+%
Linear Formula (H2C=CHCONH)2CH2
Infrared Spectrum Authentic
Formula Weight 154.17
Percent Purity ≥99.0%
Grade Biochemical
Packaging Plastic Bottle
Chemical Name or Material N, N';-Methylenebisacrylamide, For electrophoresis, For biochemistry
Show More Show Less
compliance-icons
Product Identifier
  • N,N'-Methylenebisacrylamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'