missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
N,N,N',N'-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N,N′,N′-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.
Solubility
Slightly soluble in cold, more in hot water.
Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
N,N,N′,N′-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.
Solubility
Slightly soluble in cold, more in hot water.
Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 48°C to 52°C |
Boiling Point | 260°C |
Quantity | 10 g |
UN Number | UN1673 |
Beilstein | 1564025 |
Sensitivity | Air and light sensitive |
Merck Index | 14,9228 |
Solubility Information | Slightly soluble in cold,more in hot water. |
Formula Weight | 164.25 |
Percent Purity | ≥98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'