missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%

Product Code. p-7043391
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11390104

Brand: Thermo Scientific Alfa Aesar L00559.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-22-1 | C10H16N2 | 164.252 g/mol

N,N,N',N'-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.

Solubility
Slightly soluble in cold, more in hot water.

Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-22-1
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.252
MDL Number MFCD00008309
InChI Key CJAOGUFAAWZWNI-UHFFFAOYSA-N
Synonym wurster's blue, wurster's reagent, n,n,n',n'-tetramethyl-p-phenylenediamine, n,n,n',n'-tetramethyl-1,4-phenylenediamine, tetramethyl-p-phenylenediamine, 1,4-bis dimethylamino benzene, p-bis dimethylamino benzene, 1,4-benzenediamine, n,n,n',n'-tetramethyl, benzene, 1,4-bis dimethylamino, unii-p4p3ac32zb
PubChem CID 7490
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 48°C to 52°C
Boiling Point 260°C
Quantity 10 g
UN Number UN1673
Beilstein 1564025
Sensitivity Air and light sensitive
Merck Index 14,9228
Solubility Information Slightly soluble in cold,more in hot water.
Formula Weight 164.25
Percent Purity ≥98%
Chemical Name or Material N,N,N',N'-Tetramethyl-p-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-p-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : N,N,N',N'-Tetramethyl-p-phenylenediamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.