missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%

58.69 € - 475.37 €

Chemical Identifiers

CAS 637-01-4
Molecular Formula C10H16N2·2HCl
Molecular Weight (g/mol) 237.17
MDL Number MFCD00012482
InChI Key FBHKTSXMTASXFJ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
PubChem CID 71561
IUPAC Name 1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
SMILES CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10710492
View Documents Promotion Details
Thermo Scientific Chemicals
420600050
5 g Glass bottle
58.69 €
5g
Please to purchase this item. Need a web account? Register with us today!
10740682
View Documents Promotion Details
Thermo Scientific Chemicals
420600250
25 g Glass bottle
193.97 €
25g
Please to purchase this item. Need a web account? Register with us today!
10572764
View Documents Promotion Details
Thermo Scientific Chemicals
420601000
100 g Glass bottle
475.37 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

637-01-4
237.17
FBHKTSXMTASXFJ-UHFFFAOYSA-N
71561
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
C10H16N2·2HCl
MFCD00012482
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride

Specifications

637-01-4
100.0
Beige to Gray
98+%
C6H4[N(CH3)2]2·2HCl
13, 74
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
FBHKTSXMTASXFJ-UHFFFAOYSA-N
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
71561
98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
98.5
220°C to 224°C
Authentic
C10H16N2·2HCl
MFCD00012482
10, 9052
Solubility in water: ca 50g/L
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
237.17
237.17
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 211-274-8

Recommended Storage : Product may darken on storage

Product Identifier
  • N,N,N`,N`-Tetramethyl-p-phenylenediamine dihydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only