missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetramethylbenzidine, 97.5%, Thermo Scientific Chemicals
96.58 € - 336.68 €
Chemical Identifiers
CAS | 366-29-0 |
---|---|
Molecular Formula | C16H20N2 |
Molecular Weight (g/mol) | 240.35 |
MDL Number | MFCD00008310 |
InChI Key | YRNWIFYIFSBPAU-UHFFFAOYSA-N |
Synonym | n,n,n',n'-tetramethylbenzidine, benzidine, n,n,n',n'-tetramethyl, 4,4'-bis n,n-dimethylamino biphenyl, n,n,n',n'-tetramethyl-p,p'-benzidine, ccris 1000, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 4-4-dimethylamino phenyl phenyl dimethylamine, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetramethyl |
PubChem CID | 9702 |
IUPAC Name | 4-[4-(dimethylamino)phenyl]-N,N-dimethylaniline |
SMILES | CN(C)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10795801
|
Thermo Scientific Acros
138380050 |
5 g | Glass bottle |
96.58 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10264900
|
Thermo Scientific Acros
138380250 |
25 g | Glass bottle |
336.68 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
366-29-0 | |
240.35 | |
YRNWIFYIFSBPAU-UHFFFAOYSA-N | |
9702 | |
CN(C)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C |
C16H20N2 | |
MFCD00008310 | |
n,n,n',n'-tetramethylbenzidine, benzidine, n,n,n',n'-tetramethyl, 4,4'-bis n,n-dimethylamino biphenyl, n,n,n',n'-tetramethyl-p,p'-benzidine, ccris 1000, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 4-4-dimethylamino phenyl phenyl dimethylamine, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetramethyl | |
4-[4-(dimethylamino)phenyl]-N,N-dimethylaniline |
Specifications
366-29-0 | |
100.0 | |
Brown | |
97.50% | |
((CH3)2NC6H4-)2 | |
13, 221 | |
YRNWIFYIFSBPAU-UHFFFAOYSA-N | |
4-[4-(dimethylamino)phenyl]-N,N-dimethylaniline | |
9702 | |
97.50% | |
N,N,N',N'-Tetramethylbenzidine, 97.5% |
96.5 | |
192°C to 196°C | |
Authentic | |
C16H20N2 | |
MFCD00008310 | |
n,n,n',n'-tetramethylbenzidine, benzidine, n,n,n',n'-tetramethyl, 4,4'-bis n,n-dimethylamino biphenyl, n,n,n',n'-tetramethyl-p,p'-benzidine, ccris 1000, 1,1'-biphenyl-4,4'-diamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,1'-biphenyl-4,4'-diamine, 4-4-dimethylamino phenyl phenyl dimethylamine, 1,1'-biphenyl-4,4'-diamine, n4,n4,n4',n4'-tetramethyl | |
CN(C)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C | |
240.35 | |
240.35 | |
Fine Powder |
Safety and Handling
missing translation for 'einecsNumber' : 206-676-5
missing translation for 'rtecsNumber' : DD1752100
missing translation for 'tsca' : TSCA
Product Identifier
- N,N,N',N'-Tetramethylbenzidine
RUO – Research Use Only