missing translation for 'onlineSavingsMsg'
Learn More
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure, Thermo Scientific Chemicals Product Code.: 10265090

N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure, Thermo Scientific Chemicals

Product Code. 10265090
50 mL, Glass bottle
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10265090

Brand: Thermo Scientific Acros 138450500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Melting Point -55.0°C
Density 0.7700g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH2N(CH3)2
Refractive Index 1.4169 to 1.4189
Quantity 50 mL
Beilstein 04,25
Fieser 02,403; 03,284; 04,485; 05,652; 06,576; 07,358; 11,340; 12,477; 14,68; 15,63; 16,55
Merck Index 15,9277
Specific Gravity 0.77
Solubility Information Solubility in water: miscible. Other solubilities: most organic solvents
Formula Weight 116.21
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N,N',N'-Tetramethylethylenediamine, 99%, extra pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.