missing translation for 'onlineSavingsMsg'
Learn More
N,N,N',N'-Tetramethylethylenediamine, 99%, Thermo Scientific Chemicals Product Code.: 11434214

N,N,N',N'-Tetramethylethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 11434214
500 mL
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy

Product Code. 11434214

Brand: Thermo Scientific Alfa Aesar A12536.AP

This item is not returnable. View return policy

N,N,N',N'-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylethylenediamine is used with ammonium persulfate to catalyze the polymerization of acrylamide and bis-acrylamide to get polyacrylamide gels. It is also used for the separation of proteins or nucleic acids. Further, it is employed as a ligand for metal ions like zinc and copper. It is actively involved in the formation of anionic organometallic complex.

Solubility
Miscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, carbon dioxide and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-18-9
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008335
InChI Key KWYHDKDOAIKMQN-UHFFFAOYSA-N
Synonym temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethylShow MoreShow Less
PubChem CID 8037
ChEBI CHEBI:32850
IUPAC Name N,N,N',N'-tetramethylethane-1,2-diamine
SMILES CN(C)CCN(C)C

Specifications

Melting Point -55°C
Density 0.775
Boiling Point 120°C to 122°C
Flash Point 18°C (62°F)
Assay Percent Range 99%
Odor Unpleasant
Linear Formula (CH3)2NCH2CH2N(CH3)2
Refractive Index 1.418
Quantity 500 mL
UN Number UN2372
Beilstein 1732991
Sensitivity Air sensitive; Hygroscopic
Solubility Information Miscible with water.
Formula Weight 116.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethylethylenediamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253214 Certificate TypeCertificate of Analysis Date28/11/2024 Catalog NumberA12536.0F,A12536.AE,A12536.AP
Lot Number10252345 Certificate TypeCertificate of Analysis Date09/09/2024 Catalog NumberA12536.0F,A12536.AE,A12536.AP
Lot Number10250214 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA12536.0F,A12536.AE,A12536.AP
Lot Number10249841 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog NumberA12536.0F,A12536.AE,A12536.AP
Lot Number10241137 Certificate TypeCertificate of Analysis Date20/12/2023 Catalog NumberA12536.AP
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1,2-Bis(dimethylamino)ethane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N,N,N',N'-Tetramethylethylenediamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.