missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethylmethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 11336766
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N,N,N',N'-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N,N′,N′-Tetramethylmethylenediamine is a reagent used as a convenient source of the Mannich intermediate.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-80-9
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008328
InChI Key VGIVLIHKENZQHQ-UHFFFAOYSA-N
Synonym n,n,n',n'-tetramethyldiaminomethane, bis dimethylamino methane, methanediamine, n,n,n',n'-tetramethyl, tetramethylmethylenediamine, methylenebis dimethylamine, n,n,n',n'-tetramethylmethylenediamine, unii-z870i525ks, methylenediamine, n,n,n',n'-tetramethyl, dimethylamino methyl dimethylamine, dimethyl dimethylamino methyl amine
PubChem CID 5829
SMILES CN(C)CN(C)C

Specifications

Melting Point -55°C
Density 0.749
Boiling Point 83°C to 85°C
Flash Point −12°C (10°F)
Linear Formula (CH3)2NCH2N(CH3)2
Refractive Index 1.401
Quantity 25 g
UN Number UN2733
Beilstein 1731946
Solubility Information Fully miscible in water.
IUPAC Name N,N,N',N'-tetramethylmethanediamine
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material N,N,N',N'-Tetramethylmethylenediamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'