Learn More
n-Octadecyltrichlorosilane, 95%, cont. 5-10% branched isomers, Thermo Scientific Chemicals
37.19 € - 371.85 €
Chemical Identifiers
CAS | 112-04-9 |
---|---|
Molecular Formula | C18H37Cl3Si |
Molecular Weight (g/mol) | 387.929 |
MDL Number | MFCD00000484 |
InChI Key | PYJJCSYBSYXGQQ-UHFFFAOYSA-N |
Synonym | octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane |
PubChem CID | 8157 |
IUPAC Name | trichloro(octadecyl)silane |
SMILES | CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11452368
|
Thermo Scientific Alfa Aesar
A15732.18 |
50 g |
37.19 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11462368
|
Thermo Scientific Alfa Aesar
A15732.30 |
250 g |
105.53 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442368
|
Thermo Scientific Alfa Aesar
A15732.0B |
1000 g |
371.85 €
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Octadecyltrichlorosilane is used in semiconductor industry to form self-assembled monolayer thin films on silicon dioxide substrates. Used in molecular electronics, as thin insulating gates in Metal-Insulator-Semiconductor Field-Effect Transistors. It is also used in platinum catalyzed synthesis.
Solubility
Decomposes in water.
Notes
Moisture sensitive. Store in cool dry place in tightly closed container. Store under dry inert gas. Keep away from water/moisture.
Chemical Identifiers
112-04-9 | |
387.929 | |
PYJJCSYBSYXGQQ-UHFFFAOYSA-N | |
8157 | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl |
C18H37Cl3Si | |
MFCD00000484 | |
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane | |
trichloro(octadecyl)silane |
Specifications
160°C to 162°C (3mmHg) | |
112-04-9 | |
CH3(CH2)17SiCl3 | |
UN1800 | |
189°C (372°F) | |
Decomposes in water. | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl | |
387.929 | |
387.94 | |
Pungent | |
Moisture sensitive | |
cont. 5 to 10% branched isomers |
∼20°C | |
C18H37Cl3Si | |
MFCD00000484 | |
1778847 | |
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane | |
PYJJCSYBSYXGQQ-UHFFFAOYSA-N | |
trichloro(octadecyl)silane | |
8157 | |
95% | |
1.46 | |
0.984 | |
n-Octadecyltrichlorosilane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: OCTADECYLTRICHLOROSILANE
missing translation for 'einecsNumber' : 203-930-7
missing translation for 'rtecsNumber' : VV4680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only