missing translation for 'onlineSavingsMsg'
Learn More

n-Octane, 98+%

Código de producto. 11404584
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Tamaño de la unidad:
100mL
2500mL
500mL
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto está sujeto a un impuesto especial sobre los hidrocarburos de acuerdo con la Directiva Europea 2003/96/CE.Lamentamos informarle que este producto no está disponible para Canarias, Ceuta y Melilla. Disculpe las molestias.

Código de producto. 11404584

Marca: Thermo Scientific Alfa Aesar A13181.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto está sujeto a un impuesto especial sobre los hidrocarburos de acuerdo con la Directiva Europea 2003/96/CE.Lamentamos informarle que este producto no está disponible para Canarias, Ceuta y Melilla. Disculpe las molestias.

CAS: 111-65-9 | C8H18 | 114.232 g/mol

N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-octane is mainly used as a solvent, fuel and as one of the standard values in octane rating scale. It is also used to prevent uneven burning of fuel in internal-combustion engines. Further, it acts as an antiknock agent for high compression engine fuels, lacquer diluents, blowing agent for foam rubber. In addition, it is used in rocket propellants and in azeotropic distillations.

Solubility
Miscible with ethyl ether, ethanol, acetone, benzene, petroleum ether and gasoline. Slightly Miscible with alcohol. Immiscible with water.

Notes
Hygroscopic. Incompatible with oxidizing agents and acids
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Especificaciones

Melting Point -57°C
Density 0.703
Boiling Point 125°C to 126°C
Flash Point 15°C (59°F)
Assay Percent Range ≥98%
Refractive Index 1.398
Quantity 500 mL
UN Number UN1262
Beilstein 1696875
Merck Index 14,6749
Solubility Information Miscible with ethyl ether,ethanol,acetone,benzene,petroleum ether and gasoline; Slightly Miscible with alcohol; Immiscible with water.
Formula Weight 114.23
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material n-Octane
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • n-Octane
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Otros peligros
  • MIXTURE LIST-Contains : Octane

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado