missing translation for 'onlineSavingsMsg'
Learn More

n-Octane, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Product Code. 13188633
Click to view available options
Quantity:
100 mL
1 L
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-65-9
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number MFCD00009556
InChI Key TVMXDCGIABBOFY-UHFFFAOYSA-N
Synonym n-octane, oktan, oktanen, ottani, oktanen dutch, heptane, methyl, oktan polish, ottani italian, octane, all isomers, unii-x1rv0b2fjv
PubChem CID 356
ChEBI CHEBI:17590
IUPAC Name octane
SMILES CCCCCCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -57.0°C
Color Colorless
Density 0.7080g/mL
Boiling Point 125.0°C to 127.0°C
Flash Point 13°C
Assay Percent Range 99% min. (GC)
Packaging AcroSeal glass Bottle
Linear Formula CH3(CH2)6CH3
Refractive Index 1.3965 to 1.3985
Quantity 100 mL
UN Number 1262
Residue after Evaporation 0.0003% max.
Merck Index 15, 6838
Specific Gravity 0.708
Solubility Information Solubility in water: 0.0007g/L (20°C). Other solubilities: soluble in alcohol and acetone, freely miscible with ether, benzene, petroleum, ether and gasoline
Viscosity 0.55 mPa.s (20°C)
Formula Weight 114.23
Percent Purity 99+%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material n-Octane, Extra Dry
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Octane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'