Learn More
Melford n-Octylglucoside
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Brand:  Melford N02007-25.0
 View more versions of this product
Product Code. 30018550
  • 1390.00 € / 25g
Explore more special offers
Description
Description
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Specifications
Specifications
29836-26-8
C14H28O6
25 g