missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals NG-Monomethyl-L-arginine monoacetate, 99%

Product Code. 10016213
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53308-83-1
Molecular Formula C9H20N4O4
Molecular Weight (g/mol) 248.283
MDL Number MFCD00069311
InChI Key IKPNWIGTWUZCKM-JEDNCBNOSA-N
Synonym tilarginine acetate, l-nmma, l-nmma acetate, ng-monomethyl-l-arginine acetate, nomega-me-l-arg, unii-2fl3530af2, ng-me-l-arg, acoh, tilarginine acetate usan, ng-methyl-l-arginine acetate salt, l-ornithine, n5-imino methylamino methyl-cas
PubChem CID 135242
IUPAC Name acetic acid;(2S)-2-amino-5-[(N'-methylcarbamimidoyl)amino]pentanoic acid
SMILES CC(=O)O.CN=C(N)NCCCC(C(=O)O)N

Specifications

Melting Point 180°C to 190°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Specific Rotation +9° to +13° (20°C, 589nm) (c=4, water)
Merck Index 15, 6096
Specific Rotation Condition + 10.50 (20.00°C c=4,water)
Quantity 25 mg
Formula Weight 248.3
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material N-ω-Monomethyl-L-arginine monoacetate
Show More Show Less
Product Identifier
  • N-omega-Monomethyl-L-arginine monoacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.