Learn More
n-Pentane, +99%, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -130.0°C |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Density | 0.6260g/mL |
Boiling Point | 36.0°C |
Quantity | 1 L |
Flash Point | −49°C (120.2°F) |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.358 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2404917 | Certificate of Analysis | 31/01/2025 | 170070250,170070050,170070010,170070025 |
2400125 | Certificate of Analysis | 31/01/2025 | 170070250,170070050,170070010,170070025 |
2409101 | Certificate of Analysis | 30/01/2025 | 170070250,170070050,170070010,170070025 |
1993205 | Certificate of Analysis | 27/01/2025 | 170070250,170070050,170070010,170070025 |
1997072 | Certificate of Analysis | 27/01/2025 | 170070250,170070050,170070010,170070025 |
Safety and Handling




- n-Pentane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.