missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Pentane, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -130.0°C |
Density | 0.6260g/mL |
Boiling Point | 36.0°C |
Quantity | 100 mL |
Flash Point | −49°C |
Refractive Index | 1.358 |
Specific Gravity | 0.626 |
Linear Formula | CH3(CH2)3CH3 |
Viscosity | 0.25 mPa.s (20°C) |
Water | 0.005% max. |
Show More |
Safety and Handling
Product Identifier
- n-Pentane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction