missing translation for 'onlineSavingsMsg'
Learn More

N-Pentane, Certified AR for Analysis, Fisher Chemical™

Product Code. 11458433
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

C5H12, CAS Number-109-66-0, 2.5L, 36 deg.C, CHEBI:37830, Undesignated, 72.15g/mol, OFBQJSOFQDEBGM-UHFFFAOYSA-N, pentane, -130 deg.C, 9498, 72.151, Amber glass bottle, Liquid, 8003, CCCCC, 573mbar at 20 deg.C, 0.25 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Packaging Plastic Coated Glass bottle
Quantity 2.5 L
Formula Weight 72.15g/mol
Vapor Pressure 573mbar at 20°C
Viscosity 0.25 mPaS at 20°C
Vapor Density 2.5
Melting Point -130°C
Boiling Point 36°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Pentane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
  • P331-Do NOT induce vomiting.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'