missing translation for 'onlineSavingsMsg'
Learn More

n-Pentane, Environmental Grade, 98+%, Thermo Scientific Chemicals

Product Code. 15421687
Click to view available options
Quantity:
1 L
4 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Suitable For Environmental, Pesticide, and GC Analysis

n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point °C to 130°C
Color Colorless
Appearance Colorless liquid
Density 0.626
Boiling Point 36°C
Quantity 1 L
Flash Point -49°C (-57°F)
Ignition Point 260°C (500°F)
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range >98%
Vapor Pressure 554 hPa (416mm Hg) at 20°C
Solubility Information miscible with water.
Formula Weight 72.15
Grade Environmental
Chemical Name or Material n-Pentane, Environmental Grade
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'