missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-p-phenylenediamine, 98%, Thermo Scientific Chemicals

Product Code. 11448720
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Anilinoaniline is an antioxidant that has shown to cause a reduction of NOx emissions from soybean biodiesel powered DI diesel engines but has increased CO and HC emissions. It is also used to produce dyes, pharmaceuticals, and photographic chemicals and frmerly used as a hair dye.

Solubility
Soluble in water at 20°C 0.6g/L. Soluble in (10 mg/mL) ethanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-54-2
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00007850
InChI Key ATGUVEKSASEFFO-UHFFFAOYSA-N
Synonym 4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine
PubChem CID 7564
ChEBI CHEBI:59038
IUPAC Name 4-N-phenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N

Specifications

Melting Point 72°C to 76°C
Density 1.09
Boiling Point 354°C
Flash Point 193°C (379°F)
Quantity 100 g
UN Number UN3077
Beilstein 908935
Solubility Information Soluble in water at 20°C 0.6g/L. Soluble in (10mg/mL) ethanol.
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material N-Phenyl-p-phenylenediamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'