missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylglycinonitrile, 97%, Thermo Scientific™

Product Code. 10387550
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 3009-97-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 40.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8N2
Linear Formula C6H5NHCH2CN
MDL Number MFCD00044430
Quantity 10 g
Synonym 2-phenylamino acetonitrile, n-phenylglycinonitrile, anilinoacetonitrile, n-phenylglycinenitrile, phenylamino acetonitrile, glycinonitrile, n-phenyl, phenylglycine nitrile, 1-anilinoacetonitrile, n-cyanomethyl aniline, acetonitrile, anilino
InChI Key KAXCEFLQAYFJKV-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NCC#N
IUPAC Name 2-anilinoacetonitrile
Molecular Weight (g/mol) 132.16
PubChem CID 76372
Formula Weight 132.16
Percent Purity 97%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material N-Phenylglycinonitrile, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Phenylglycinonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.