missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylthiourea, 97%, Thermo Scientific Chemicals

Product Code. 11496730
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

Solubility
Soluble in hot water and alcohol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES NC(=S)NC1=CC=CC=C1

Specifications

Melting Point 149°C to 153°C
Odor Odorless
Quantity 50 g
UN Number UN2811
Beilstein 907309
Merck Index 14,7315
Solubility Information Soluble in hot water and alcohol.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material N-Phenylthiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'