missing translation for 'onlineSavingsMsg'
Learn More

N-Propylethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 10195850
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-39-7
Molecular Formula C5H14N2
Molecular Weight (g/mol) 102.18
MDL Number MFCD00008172
InChI Key CFNHVUGPXZUTRR-UHFFFAOYSA-N
Synonym n-propylethylenediamine, n-n-propyl ethylenediamine, 1,2-ethanediamine, n-propyl, ethylenediamine, n-propyl, 2-aminoethyl propyl amine, 1,2-ethanediamine, n1-propyl, 2-n-propylamino ethylamine, 2-n-propylaminoethylamine, propylethylenediamine, n-n-propylethylenediamine
PubChem CID 66073
IUPAC Name N'-propylethane-1,2-diamine
SMILES CCCNCCN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8200g/mL
Boiling Point 147°C to 150°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3CH2CH2NHCH2CH2NH2
Refractive Index 1.4394 to 1.4414
Quantity 5 g
Specific Gravity 0.82
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Propylethylenediamine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'