missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl 3-maleimidobenzoate, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates
Solubility
Soluble in DMF or ethyl acetate.
Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates
Solubility
Soluble in DMF or ethyl acetate.
Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 175°C to 177°C |
Color | White |
Quantity | 1 g |
Beilstein | 1505254 |
Sensitivity | Light and moisture sensitive |
Solubility Information | Soluble in DMF or ethyl acetate. |
Formula Weight | 314.26 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | N-Succinimidyl 3-maleimidobenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'