missing translation for 'onlineSavingsMsg'
Learn More
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide, 97%, Thermo Scientific™ Product Code.: 11975671

N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide, 97%, Thermo Scientific™

Product Code. 11975671
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11975671

Brand: Thermo Scientific Acros 440580050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 87694-52-8
Molecular Formula C12H24N2O4
Molecular Weight (g/mol) 260.33
MDL Number MFCD00151897
InChI Key RRBFCGUIFHFYQK-VIFPVBQESA-N
Synonym s-2-boc-amino-n-methoxy-n,3-dimethylbutanamide, n-boc-l-valine n'-methoxy-n'-methylamide, n-tert-butoxycarbonyl-l-valine n'-methoxy-n'-methylamide, tert-butyl s-1-methoxy methyl amino-3-methyl-1-oxobutan-2-yl carbamate, tert-butyl n-1s-1-methoxy methyl carbamoyl-2-methylpropyl carbamate, boc-val-n och3 ch3, n-boc-l-valinen'-methoxy-n'-methylamide, n-alpha-t-butyloxycarbonyl-n-methyl-n-methoxy-l-valine amide, 2s-2-tert-butoxy carbonylamino-n-methoxy-3-methyl-n-methylbutanamideShow MoreShow Less
PubChem CID 10083961
IUPAC Name tert-butyl N-[(2S)-1-[methoxy(methyl)amino]-3-methyl-1-oxobutan-2-yl]carbamate
SMILES CON(C)C(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Color White to Yellow
Boiling Point 248°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Rotation -15
Formula Weight 260.34
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide
Show More Show Less
Product Identifier
  • N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.