missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 105°C to 110°C |
Density | 1.29 |
Flash Point | 165°C (329°F) |
Quantity | 50 g |
Beilstein | 158370 |
Formula Weight | 238.38 |
Percent Purity | 97% |
Chemical Name or Material | N-tert-Butyl-2-benzothiazolesulfenamide |
Safety and Handling


Product Identifier
- N-tert-Butyl-2-benzothiazolesulfenamide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : N-tert-Butyl-2-benzothiazolesulfenamide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
N-tert-Butyl-2-benzothiazolesulfenamide, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction