missing translation for 'onlineSavingsMsg'
Learn More
N-(tert-Butyl)hydroxylamine hydrochloride, 98% Product Code.: 10768425

N-(tert-Butyl)hydroxylamine hydrochloride, 98%

Product Code. 10768425
1 g, Glass bottle
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10768425

Brand: Thermo Scientific Acros 186910010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57497-39-9 | C4H11NO·HCl | 125.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 57497-39-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 176.0°C to 180.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C4H11NO·HCl
Linear Formula (CH3)3CNHOH·HCl
MDL Number MFCD00012598
Quantity 1 g
Fieser 12,95; 04,191; 05,47; 06,64; 08,49; 10,49; 14,52; 17,50
Synonym n-tert-butyl hydroxylamine hydrochloride, n-tert-butyl hydroxylamine hcl, n-tert-butylhydroxylamine hydrochloride, n-1,1-dimethylethyl hydroxylamine hydrochloride, 2-propanamine, n-hydroxy-2-methyl-, hydrochloride, 2-methylprop-2-ylhydroxylamine, chloride, soyheadhjuup@, acmc-1b1z0, t-butylhydroxylamine hydrochloride
InChI Key DCSATTBHEMKGIP-UHFFFAOYSA-N
SMILES CC(C)(C)NO.Cl
IUPAC Name N-tert-butylhydroxylamine;hydrochloride
Molecular Weight (g/mol) 125.6
PubChem CID 2777896
Formula Weight 125.6
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-(tert-Butyl)hydroxylamine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0371846 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number186910010
compliance-icons
Product Identifier
  • N-(tert-Butyl)hydroxylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(tert-Butyl)hydroxylamine hydrochloride, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.