missing translation for 'onlineSavingsMsg'
Learn More
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide, 97%, Thermo Scientific Chemicals Product Code.: 11420208

N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide, 97%, Thermo Scientific Chemicals

Product Code. 11420208
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11420208

Brand: Thermo Scientific Alfa Aesar B23760.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It finds its application in the preparation of N-tert-butyldimethylsilyl ethanolamines resulting from hydrolysis of nitrogen mustards and a silylating agent used in the derivatization of various organic compounds (such as amino acids) for gas chromatography-mass spectrometry (GC-MS) analysis and selective O-silylation of N,O-diacylhydroxylamines and GLC derivatization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the preparation of N-tert-butyldimethylsilyl ethanolamines resulting from hydrolysis of nitrogen mustards and a silylating agent used in the derivatization of various organic compounds (such as amino acids) for gas chromatography-mass spectrometry (GC-MS) analysis and selective O-silylation of N,O-diacylhydroxylamines and GLC derivatization.

Solubility
Soluble in water (reacts).

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store away from electrostatic charges. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77377-52-7
Molecular Formula C9H18F3NOSi
Molecular Weight (g/mol) 241.329
MDL Number MFCD00009671
InChI Key QRKUHYFDBWGLHJ-UHFFFAOYSA-N
Synonym mtbstfa, n-tert-butyldimethylsilyl-n-methyltrifluoroacetamide, n-methyl-n-tert-butyldimethylsilyl trifluoroacetamide, n-tert-butyldimethylsilyl-n-methyl-trifluoroacetamide, n-methyl-n-t-butyldimethylsilyl trifluoroacetamide, acetamide, n-1,1-dimethylethyl dimethylsilyl-2,2,2-trifluoro-n-methyl, n-t-butyldimethylsilyl-n-methyltrifluoroacetamide, n-t-butyldimethylsilyl-n-methyltrifluoro-acetamideShow MoreShow Less
PubChem CID 2724275
ChEBI CHEBI:85060
IUPAC Name N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide
SMILES CC(C)(C)[Si](C)(C)N(C)C(=O)C(F)(F)F

Specifications

Density 1.023
Boiling Point 168°C to 170°C
Flash Point 52°C (125°F)
Refractive Index 1.402
Quantity 25 g
UN Number UN1993
Beilstein 3606546
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts).
Formula Weight 241.33
Percent Purity 97%
Chemical Name or Material N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10160886 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23760.06,B23760.03,B23760.14
Lot Number10204075 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23760.06,B23760.03,B23760.14
Lot Number10156706 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23760.06,B23760.03,B23760.14
Lot Number10177997 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23760.06,B23760.03,B23760.14
Lot Number10215295 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23760.06,B23760.03,B23760.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.