missing translation for 'onlineSavingsMsg'
Learn More

n-Undecanoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 15432848
Click to view available options
Quantity:
500 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17746-05-3
Molecular Formula C11H21ClO
Molecular Weight (g/mol) 204.738
MDL Number MFCD00000739
InChI Key JUKPJGZUFHCZQI-UHFFFAOYSA-N
Synonym n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
PubChem CID 87287
IUPAC Name undecanoyl chloride
SMILES CCCCCCCCCCC(=O)Cl

Specifications

Boiling Point 135°C to 136°C
Flash Point 113°C (235°F)
Linear Formula CH3(CH2)9COCl
Refractive Index 1.443
Quantity 1 g
UN Number UN3265
Beilstein 1759226
Sensitivity Moisture sensitive
Formula Weight 204.74
Percent Purity 98%
Chemical Name or Material n-Undecanoyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'