Learn More
N-Vinyl-2-pyrrolidone, 99%, stabilized with NaOH, Thermo Scientific Chemicals
68.04 € - 1225.00 €
Chemical Identifiers
CAS | 88-12-0 |
---|---|
Molecular Formula | C6H9NO |
Molecular Weight (g/mol) | 111.14 |
MDL Number | MFCD00003197 |
InChI Key | WHNWPMSKXPGLAX-UHFFFAOYSA-N |
Synonym | n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone |
PubChem CID | 6917 |
ChEBI | CHEBI:82551 |
IUPAC Name | 1-ethenylpyrrolidin-2-one |
SMILES | C=CN1CCCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10071500
|
Thermo Scientific Acros
140920050 |
5 mL |
68.04 €
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10193090
|
Thermo Scientific Acros
140925000 |
500 mL |
120.60 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
|||||||||
10773694
|
Thermo Scientific Acros
140920025 |
2.5 L |
399.99 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15264634
|
Thermo Scientific Acros
140920100 |
10 L | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
88-12-0 | |
111.14 | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
6917 | |
1-ethenylpyrrolidin-2-one |
C6H9NO | |
MFCD00003197 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
CHEBI:82551 | |
C=CN1CCCC1=O |
Specifications
88-12-0 | |
100.0 | |
Colorless to Yellow | |
218°C | |
Authentic | |
Glass bottle | |
1.5119 to 1.5139 | |
15,7814 | |
n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone | |
WHNWPMSKXPGLAX-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one | |
111.14 | |
CHEBI:82551 | |
111.14 | |
Liquid |
98.5 | |
13°C to 14°C | |
1.0430g/mL | |
95°C | |
0.2% max. (Pyrrolidone) (GC) | |
C6H9NO | |
MFCD00003197 | |
1.043 | |
Solubility in water: 52.1g/L (25°C). Other solubilities: miscible with acetone,diethyl ether,ethanol,,toluene,benzene,very good soluble in organic solvents | |
C=CN1CCCC1=O | |
0.1% flaked caustic soda | |
6917 | |
2.4 mPa.s (20°C) | |
99% | |
N-Vinyl-2-pyrrolidone, 98% |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-800-4
missing translation for 'rtecsNumber' : UY6107000
missing translation for 'tsca' : TSCA
Recommended Storage : Product may darken on storage
- N-Vinyl-2-pyrrolidone, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.