missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1-(4-Aminophenyl)-N1-methylacetamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 164.21 |
Percent Purity | Tech.. |
Grade | Technical |
Chemical Name or Material | N1-(4-Aminophenyl)-N1-methylacetamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'