missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1-(4-Butyryl-3-hydroxyphenyl)acetamide, Tech., Thermo Scientific™
58.00 € - 151.00 €
Identificatori chimici
CAS | 28583-62-2 |
---|---|
Molecular Formula | C12H15NO3 |
Molecular Weight (g/mol) | 221.26 |
MDL Number | MFCD00276573 |
InChI Key | RSBIKRQPSYDRFZ-UHFFFAOYSA-N |
Synonym | n1-4-butyryl-3-hydroxyphenyl acetamide, unii-4kn634aqj7, n-4-butanoyl-3-hydroxyphenyl acetamide, 4'-butyryl-3'-hydroxyacetanilide, n-4-butyryl-3-hydroxyphenyl acetamide, n-3-hydroxy-4-1-oxobutyl phenyl acetamide, maybridge3_006119, acetamide,n-3-hydroxy-4-1-oxobutyl phenyl, acetamide, n-3-hydroxy-4-1-oxobutyl phenyl |
PubChem CID | 119989 |
IUPAC Name | N-(4-butanoyl-3-hydroxyphenyl)acetamide |
SMILES | CCCC(=O)C1=CC=C(NC(C)=O)C=C1O |
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
10495132
|
Thermo Scientific Maybridge
SB01739DA |
1 g |
58.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10006374
|
Thermo Scientific Maybridge
SB01739DE |
5 g |
93.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10049571
|
Thermo Scientific Maybridge
SB01739EA |
10 g |
151.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
Identificatori chimici
28583-62-2 | |
221.26 | |
RSBIKRQPSYDRFZ-UHFFFAOYSA-N | |
119989 | |
CCCC(=O)C1=CC=C(NC(C)=O)C=C1O |
C12H15NO3 | |
MFCD00276573 | |
n1-4-butyryl-3-hydroxyphenyl acetamide, unii-4kn634aqj7, n-4-butanoyl-3-hydroxyphenyl acetamide, 4'-butyryl-3'-hydroxyacetanilide, n-4-butyryl-3-hydroxyphenyl acetamide, n-3-hydroxy-4-1-oxobutyl phenyl acetamide, maybridge3_006119, acetamide,n-3-hydroxy-4-1-oxobutyl phenyl, acetamide, n-3-hydroxy-4-1-oxobutyl phenyl | |
N-(4-butanoyl-3-hydroxyphenyl)acetamide |
Specifica
28583-62-2 | |
C12H15NO3 | |
n1-4-butyryl-3-hydroxyphenyl acetamide, unii-4kn634aqj7, n-4-butanoyl-3-hydroxyphenyl acetamide, 4'-butyryl-3'-hydroxyacetanilide, n-4-butyryl-3-hydroxyphenyl acetamide, n-3-hydroxy-4-1-oxobutyl phenyl acetamide, maybridge3_006119, acetamide,n-3-hydroxy-4-1-oxobutyl phenyl, acetamide, n-3-hydroxy-4-1-oxobutyl phenyl | |
CCCC(=O)C1=CC=C(NC(C)=O)C=C1O | |
221.26 | |
221.25 | |
Technical |
Amber glass bottle | |
MFCD00276573 | |
RSBIKRQPSYDRFZ-UHFFFAOYSA-N | |
N-(4-butanoyl-3-hydroxyphenyl)acetamide | |
119989 | |
Tech. | |
N1-(4-Butyryl-3-hydroxyphenyl)acetamide |