Learn More
N1-Methyl-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™
27.40 € - 283.00 €
Chemical Identifiers
CAS | 41419-59-4 |
---|---|
Molecular Formula | C8H8F3NO |
Molecular Weight (g/mol) | 191.153 |
MDL Number | MFCD00052332 |
InChI Key | MGCCWCLGIPNIBP-UHFFFAOYSA-N |
Synonym | n-methyl-4-trifluoromethoxy aniline, n1-methyl-4-trifluoromethoxy aniline, benzenamine,n-methyl-4-trifluoromethoxy, methyl 4-trifluoromethoxy phenyl amine, pubchem8511, 4-trifluoromethoxy-n-methylaniline, methyl 4-trifluoromethoxyphenyl amine, methyl-4-trifluoromethoxy-phenyl-amine, n-methyl-4-trifluoromethoxyphenyl amine |
PubChem CID | 737351 |
IUPAC Name | N-methyl-4-(trifluoromethoxy)aniline |
SMILES | CNC1=CC=C(C=C1)OC(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10079441
|
Thermo Scientific Maybridge
TL00405DA |
1 g |
27.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10486862
|
Thermo Scientific Maybridge
TL00405EA |
10 g |
145.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10016394
|
Thermo Scientific Maybridge
TL00405EB |
25 g |
283.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
41419-59-4 | |
191.153 | |
MGCCWCLGIPNIBP-UHFFFAOYSA-N | |
737351 | |
CNC1=CC=C(C=C1)OC(F)(F)F |
C8H8F3NO | |
MFCD00052332 | |
n-methyl-4-trifluoromethoxy aniline, n1-methyl-4-trifluoromethoxy aniline, benzenamine,n-methyl-4-trifluoromethoxy, methyl 4-trifluoromethoxy phenyl amine, pubchem8511, 4-trifluoromethoxy-n-methylaniline, methyl 4-trifluoromethoxyphenyl amine, methyl-4-trifluoromethoxy-phenyl-amine, n-methyl-4-trifluoromethoxyphenyl amine | |
N-methyl-4-(trifluoromethoxy)aniline |
Specifications
41419-59-4 | |
95% | |
C8H8F3NO | |
MFCD00052332 | |
MGCCWCLGIPNIBP-UHFFFAOYSA-N | |
N-methyl-4-(trifluoromethoxy)aniline | |
737351 | |
95% |
95.0 | |
Amber glass bottle | |
CF3OC6H4NHCH3 | |
n-methyl-4-trifluoromethoxy aniline, n1-methyl-4-trifluoromethoxy aniline, benzenamine,n-methyl-4-trifluoromethoxy, methyl 4-trifluoromethoxy phenyl amine, pubchem8511, 4-trifluoromethoxy-n-methylaniline, methyl 4-trifluoromethoxyphenyl amine, methyl-4-trifluoromethoxy-phenyl-amine, n-methyl-4-trifluoromethoxyphenyl amine | |
CNC1=CC=C(C=C1)OC(F)(F)F | |
191.153 | |
191.15 | |
N1-methyl-4-(trifluoromethoxy)aniline |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.