Learn More
Macherey-Nagel™ N20 Aluminum Closures
Seal vials with securely with a variety of crimp closures. Machenery-Nagel™ N20 aluminum closures come with a broad variety of cap types and septum materials.
Brand: Macherey-Nagel™ 70237
This item is not returnable.
View return policy
Description
- Center tear off caps and complete tear off caps N 20
- Bi-metal crimp caps and magnetic crimp caps N 20
- for use on autosamplers with magnetic transportation system –
- Crimping of the bi-metal crimp caps is more convenient.
- All closures mentioned above are available with silicone / PTFE septa as well as with butyl / PTFE septa in different versions.
- Applications above 120 °C require silicone / PTFE septa.
- Individual septa, stoppers as well as PE caps N 20 are also available.
- Manual and electronic crimping tools N 20 complete this product range
Specifications
Silver | |
Butyl stopper, grey unassembled (separate parts) | |
Crimp Cap |
Aluminum, Butyl Rubber | |
100 Pk. |