missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methylcytidine-3'-(2-cyanoethyl diisopropylphosphoramidite), 98%

Product Code. 15484465
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15484465

Brand: Thermo Scientific Chemicals J65701.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

C47H54N5O9P

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N4-Benzoyl-5'-O-(4,4'-dimethoxytrityl)-2'-O-methylcytidine-3'-(2-cyanoethyl diisopropylphosphoramidi
Color White
Molecular Formula C47H54N5O9P
MDL Number MFCD00792624
Quantity 1 g
Formula Weight 863.95
Percent Purity 98%
Physical Form Powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.