Learn More
N4BP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16733-1-AP-150UL
 View more versions of this product
Product Code. 16826464
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This antibody detects 60 kDa N4BP3 protein and a weaker 40 kDa protein.
N4BP3, also named as KIAA0341, a Fezzin family member, during nerve cell development. It is crucial for axonal and dendritic branching in developing neurons. It binds the protein NEDD4 and interacts with 14-3-3 proteins. N4BP3 is present in early neurites, growth cones, axons and dendrites of developing primary hippocampal neurons.
Specifications
Specifications
N4BP3
Polyclonal
Unconjugated
N4BP3
KIAA0341, N4BP3, Nedd4 binding protein 3
Rabbit
Antigen Affinity Chromatography
RUO
212706, 23138, 303112
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15049, Q3LUD3, Q8C7U1
N4bp3
N4BP3 Fusion Protein Ag10194
150 μL
Primary
Human, Mouse, Rat