Learn More
Honeywell™ N7700 Half Mask Respirator Twin
N7700 high performance twin filter half-masks are lightweight with a low profile ergonomic design for a good fit and wearer comfort. The unique cradle headband and yoke piece give ease of adjustment. The contoured revert seal gives an excellent face seal that will not cause discomfort.
Brand:  Honeywell™ N65770031
Product Code. 15846845
  • 8.44 € / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • PPE Category III
  • EN 140, CE Cert 250 - 7700 Respir - Dec 2007
  • 100% silicone hypoallergenic face piece - extra comfort
  • Single unit four-point harness with neck strap - Easy to use, ergonomic design and improved suspension system fits perfectly on top of the head providing a high level of comfort
  • Reduced dead air space - less re-breathed exhaled air
  • Quick to dismantle and clean, with replaceable parts - easy maintenance
  • Superior exhalation valve allows more breath to be exhaled - workers feel cooler and more comfortable
  • Variable thickness in sealing area conforms to nose bridge - improved comfort and fit
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PPE CAT III, EN 140
Threaded Rd40
Reusable
Half Mask Respirator
Yes
Laboratories, pharmaceutical industry - exposure to aerosols containing bacteria or viruses, handling powders, cleanrooms, oil and gas industry - handling chemicals where there is no risk of splashes (gas detection).
Single unit four-point harness with neck strap
EN 140
Black
Class 1
Small
100% Silicone
Yes