missing translation for 'onlineSavingsMsg'
Learn More

NAADP tetrasodium salt, Tocris Bioscience™

Product Code. 18044662
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Ca2+ mobilizing agent

Ca2+ mobilizing agent. Initiates Ca2+ release via type 1 ryanodine receptor (RyR1) activation. Also regulates Ca2+ release from intracellular stores distinct from the endoplasmic reticulum (ER); displays affinity for two-pore channels (TPCs) which release Ca2+ from acidic organelles.

Specifications

CAS 5502-96-5
Molecular Formula C21H23N6Na4O18P3
Quantity 10 mg
Synonym naadp, naadp tetrasodium salt
InChI Key NWKIGVGTZIAOCW-MCNGMCCKSA-J
SMILES C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)([O-])[O-])O)O)O)C(=O)[O-].[Na+].[Na+].[Na+].[Na+]
IUPAC Name tetrasodium;1-[(2R,3R,4S,5R)-5-[[[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonatooxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl]oxymethyl]-3,4-dihydroxyoxolan-2-yl]pyridin-1-ium-3-carboxylate
Molecular Weight (g/mol) 832.319
PubChem CID 90488897
Formula Weight 832.32
Percent Purity >98%
Chemical Name or Material NAADP tetrasodium salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'