missing translation for 'onlineSavingsMsg'
Learn More

Nafion∣r N-115 membrane, 0.125mm thick, ∣>0.90 meq/g exchange capacity, Thermo Scientific Chemicals

Product Code. 11349827
Click to view available options
Quantity:
15 x 15 cm
30 x 30 cm
60 x 60 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Widely used for proton exchange membrane (PEM) fuel cells and water electrolyzers. It is a popular separator in electrochemical cells requiring high ionic conductivity such as fuel cells and water electrolyzers.

Nafion™ N-115 membrane is widely used for proton exchange membrane (PEM) fuel cells and water electrolyzers. It is a popular separator in electrochemical cells requiring high ionic conductivity such as fuel cells and water electrolyzers. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31175-20-9
Molecular Formula C9HF17O5S
Molecular Weight (g/mol) 544.135
MDL Number MFCD00132790
InChI Key FOYUGSIADQEOEK-UHFFFAOYSA-N
Synonym nafion, nafion nr 50 beads 10-35 mesh, perfluoro 2-2-sulfonylethoxy propyl vinyl ether-tetrafluoroethylene copolymer, tetrafluoroethylene-perfluoro 2-2-sulfonylethoxy propyl vinyl ether copolymer, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoroethanesulfonyl fluoride-tetrafl, ethanesulfonic acid, 1,1,2,2-tetrafluoro-2-1,2,2-trifluoro-1-trifluoromethyl-2-1,2,2-trifluorovinyl oxy ethoxy-, pol, ethanesulfonic acid, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer, nafion n-324 membrane, 0.15mm 0.006in thick, teflon fabric reinforced, nafion nre-212 membrane, 0.05mm thick, ≥0.92 meq/g exchange capacity, nafion d-520 dispersion w/w in water and 1-propanol, ≥1.00 meq/g exchange capacity
PubChem CID 61889
IUPAC Name 1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonic acid
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)O)(F)F)F)(F)F)F.C(=C(F)F)(F)F

Specifications

Density 1.98
Packaging Shrink-wrapped
Quantity 15 x 15 cm
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Chemical Name or Material Nafion™ N-115 membrane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'