missing translation for 'onlineSavingsMsg'
Learn More

Nafion∣r N-324 membrane, 0.15mm (0.006in) thick, Teflon∣r fabric reinforced, Thermo Scientific Chemicals

Product Code. 15437105
Click to view available options
Quantity:
15 x 15 cm
30 x 30 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

Nafion™ N-324 membrane is used in chloroalkali production and a variety of electrolytic processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31175-20-9
Molecular Formula C9HF17O5S
Molecular Weight (g/mol) 544.135
MDL Number MFCD00132790
InChI Key FOYUGSIADQEOEK-UHFFFAOYSA-N
Synonym nafion, nafion nr 50 beads 10-35 mesh, perfluoro 2-2-sulfonylethoxy propyl vinyl ether-tetrafluoroethylene copolymer, tetrafluoroethylene-perfluoro 2-2-sulfonylethoxy propyl vinyl ether copolymer, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoroethanesulfonyl fluoride-tetrafl, ethanesulfonic acid, 1,1,2,2-tetrafluoro-2-1,2,2-trifluoro-1-trifluoromethyl-2-1,2,2-trifluorovinyl oxy ethoxy-, pol, ethanesulfonic acid, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer, nafion n-324 membrane, 0.15mm 0.006in thick, teflon fabric reinforced, nafion nre-212 membrane, 0.05mm thick, ≥0.92 meq/g exchange capacity, nafion d-520 dispersion w/w in water and 1-propanol, ≥1.00 meq/g exchange capacity
PubChem CID 61889
IUPAC Name 1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonic acid
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)O)(F)F)F)(F)F)F.C(=C(F)F)(F)F

Specifications

Quantity 15 x 15 cm
Solubility Information Insoluble in water.
Chemical Name or Material Nafion™ N-324 membrane, 0.15mm (0.006 in.) thick, PTFE fabric reinforced

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'