missing translation for 'onlineSavingsMsg'
Learn More

Nafion∣r R-1100 membrane, 0.05mm thick, sulfonyl fluoride form, Thermo Scientific Chemicals

Product Code. 15467175
Click to view available options
Quantity:
25 x 25 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Nafion is used in ion exchange applications, a perfluorinated precursor resin in the sulfonyl fluoride form.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 26654-97-7
Molecular Formula C9F18O4S
MDL Number MFCD05665455
Quantity 25 x 25 cm
Synonym 1,1,2,2-tetrafluoroethene; 1,1,2,2-tetrafluoro-2-1,1,1,2,3,3-hexafluoro-3-1,2,2-trifluoroethenoxy propan-2-yl oxyethanesulfonyl fluoride, 1,1,2,2-tetrafluoro-2-1,1,1,2,3,3-hexafluoro-3-trifluoroethenyl oxy propan-2-yl oxy ethanesulfonyl fluoride-tetrafluoroethene 1:1, tetrafluoroethylene-perfluoro sulfinoethoxypropoxytrifluoroethylene copolymer, ethanesulfonyl fluoride, 2-1-difluoro 1,2,2-trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer with 1,1,2,2-tetrafluoroethene, ethanesulfonyl fluoride, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer with tetrafluoroethene, nafion r r-1100 resin, sulfonyl fluoride form 1g, nafion r-1100 membrane, 0.05mm thick, sulfonyl fluoride form, 1,1,2,2-tetrakis fluoranyl ethene; 1,1,2,2-tetrakis fluoranyl-2-1,1,1,2,3,3-hexakis fluoranyl-3-1,2,2-tris fluoranyl ethenoxy propan-2-yl oxy-ethanesulfonyl fluoride
Solubility Information Insoluble in water.
InChI Key DHYYUWYUPUOPFL-UHFFFAOYSA-N
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)F)(F)F)F)(F)F)F.C(=C(F)F)(F)F
IUPAC Name 1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonyl fluoride
Molecular Weight (g/mol) 546.126
PubChem CID 117323
Chemical Name or Material Nafion™ R-1100 membrane, 0.05mm thick, sulfonyl fluoride form
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.