Learn More
Rogo Sampaic™ Nageotte Counting Cells
For determining the number of particles per volume unit of a liquid. Rogo Sampaic™ Nageotte Counting Cells have a simple or double grid with a depth of 0.5mm.
Specifications
Product Type Nageotte Cell
Quantity 1Each
Sample Type Spinal, Urinary
Depth (Metric) 0.5mm
View More Specs
Description
Description
  • For spinal and urinary counts
  • Provides for accurate manual measurements
Specifications
Specifications
Nageotte Cell
1Each
Spinal, Urinary
0.5mm