missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Carboy Replacement Screw Closures and Gaskets

Product Code. 10254451
Click to view available options
Closure Size:
100 mm
120 mm
53B
83B
Product Type:
Screw Cap
Screw Cap with Strap
Screw Closure and Gasket
Sealing Gasket
Material:
HDPE
Polypropylene
Silicone
TPE
This item is not returnable. View return policy
This item is not returnable. View return policy

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

Secure-fitting Thermo Scientific™ Nalgene™ Replacement Screw Closures and Gaskets for Nalgene Carboys provide leakproof† service

  • Made to fit Nalgene Carboys
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Replacement Closure
Material HDPE
No. per Case 12
No. per Pack 12
Closure Included Yes
Shape Round
Product Type Screw Closure and Gasket
For Use With (Equipment) Nalgene Large Carboys and Bottles
Closure Material HDPE
Closure Size 53B
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.