Learn More
Thermo Scientific™ Nalgene™Polycarbonate Biotainer™Bottles and Carboys, 5L
Product Code. 15656638
Specifically designed for pharmaceutical and biological manufacturing to reduce the risk of carry-over contamination. Durable, translucent PC construction ensures the integrity of your sample, even frozen to -100°C. 
Provided sterile and ready-to-use; including certified clean products, which meet USP <788> standards and endotoxins below 0.5 EU/mL. 
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™
Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys reduce the risk of carry-over
contamination. Durable, translucent PC construction ensures the integrity of your sample, even
frozen to -100°C.
  • Provided sterile and ready-to-use; including certified clean products, which meet USP<788> standards and endotoxins below 0.5 EU/mL
  • Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of  entry into cleanrooms
  • Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of entry into cleanrooms
  • Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the bottle/closure and two heat-sealed sterile barriers to protect product prior to use
  • All heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging
  • Certified to meet USP <788> low particulate requirements to limit potential particulate contamination
  • Certified low endotoxin (0.50 EU/mL) according to USP<85> to reduce pyrogens
  • Products can be supplied with forced extraction studies*, validation binders* and certificates
  • Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications
  • Clean roomproduced in ISO Class 7 to limit particulates during molding 
  • All container/closure systems are certified leakproof** to protect valuable contents
  • Robust, PC can be used from -100˚C to 100˚C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application.
  • Customer support includes our highly experienced commercial specialist teams, regulatory team, quality team and regional technical support