missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Clear PETG Diagnostic Bottles with Closure: Sterile, Bulk Pack
GREENER_CHOICE

Product Code. 10501465
Click to view available options
Capacity (Metric):
10 mL
20 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Sample, store and ship small-volume, valuable reagents with clear PETG diagnostic bottles that are ideal for use in many sterile, small-volume applications.

Sample, store and ship small-volume, valuable reagents using Thermo Scientific™ Nalgene Clear PETG Diagnostic Bottles. These bottles are ideal for use in test kits and in many sterile, small-volume applications.

Features:

  • Polyethylene Terephthalate (PETG) with excellent clarity for viewing contents
  • Non-pyrogenic and non-cytotoxic
  • Includes lined, white, high-density polyethylene (HDPE) closure for a leakproof† seal

Ordering Information:

Serum vials are packed in shrink-wrapped trays with closures attached.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.33 oz., 10 mL
Capacity (Metric) 10 mL
Capacity (English) 0.33 oz.
Diameter (Metric) Inner Neck 11 mm
Material PETG
No. per Case 500
No. per Pack 100
Sterility Sterile
Closure Color White
Neck Finish 20-415 mm
Certifications/Compliance Manufactured in compliance with ISO 13485:2016 quality requirements. Serum Vials meet the requirements of 21CFR177.1315 for food and beverage use, USP Class VI.
Closure Included Yes
Mouth Narrow
Height (Metric) 54 mm
Product Type Sample Container
Width (English) 0.93 in.
Height (English) 2.12 in.
Closure Material HDPE
Diameter (Metric) 24 mm
Through the Closure Dispensing No
Show More Show Less

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.