missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Closed-Dome Bio Tank Closure with Mixer Support Assembly

Enable overhead mixing in a closed system with this unique flange assembly that can be used with all sizes of Thermo Scientific™ Nalgene™ Closed-Dome Tanks.

Brand:  Thermo Scientific™ 2651-0200

850.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10325751

  • 877.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enable overhead mixing in a closed system with this unique, sanitary flange assembly. Thermo Scientific™ Nalgene™ Closed-Dome Bio Tank Closure with Mixer Support Assembly is for use with all sizes of Nalgene™ Closed-Dome Bio Tanks. Nalgene™ Biotech Mixer is not included.

Features:

  • 6 in. (15.2cm) polypropylene screw closure with 2 in. (5.1cm) sanitary ferrule welded in the center
  • 2 in. (5.1cm) silicone gasket and PDVF True Union clamp
  • Autoclavable

Compatible with:
Use with Nalgene Closed-Dome Tanks (Series 2650)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Closed dome tank closure w/support assembly
Bio Tank
1
152.3 mm
Polypropylene, PVDF, Silicone
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Schematics & Diagrams

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1414759 Certificate TypeCertificate of Compliance Date12/10/2024 Catalog Number2651-0200
Lot Number1402990 Certificate TypeCertificate of Compliance Date14/03/2024 Catalog Number2651-0200
Lot Number1384924 Certificate TypeCertificate of Compliance Date28/04/2023 Catalog Number2651-0200
Lot Number1374413 Certificate TypeCertificate of Compliance Date13/02/2023 Catalog Number2651-0200
Lot Number1358354 Certificate TypeCertificate of Compliance Date06/10/2022 Catalog Number2651-0200
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Closed-Dome Bio Tank Closure with Mixer Support Assembly > Closed dome tank closure w/supp

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.