missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners

Enhance mixing capabilities open-bag, flat-bottom Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners , which offer cost-effective, single-use fluid processing.

785.00 € - 1175.00 €

Specifications

Type Tank Liner
Material Polyethylene
For Use With (Equipment) Nalgene cylindrical tanks from 5 gal (19L) to 200
No. per Case 10
No. per Pack 10
View More Specs

Products 4
Product Code Brand Capacity (English) Price Quantity & Availability  
Product Code Brand Capacity (English) Price Quantity & Availability  
10561695
View Documents
Thermo Scientific™
343050-0030
30 gal.
785.00 €
Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
10767104
View Documents
Thermo Scientific™
343050-0055
55 gal.
861.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
 
10405183
View Documents
Thermo Scientific™
343050-0100
100 gal.
959.00 €
Pack of 10
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
 
10079632
View Documents
Thermo Scientific™
343050-0150
150 gal.
1175.00 €
Pack of 10
In Stock
to see stock.
Add to basket
 
Description

Description

Enhance mixing capabilities with open-bag, flat-bottom liners. Thermo Scientific™ Nalgene™ Coex Polyethylene Film Tank Liners offer cost-effective, single-use biopharmaceutical and diagnostic reagent fluid processing with rigid Nalgene cylindrical plastic tanks.

  • Designed to fit Nalgene cylindrical tanks from 5 gal (19L) to 200 gal (787L)
  • Gamma-irradiated, food-grade film is noncytotoxic and certified nonpyrogenic

Warranty: 90 days
Specifications

Specifications

Tank Liner
Nalgene cylindrical tanks from 5 gal (19L) to 200
10
Polyethylene
10
Videos
SDS
Documents

Documents

Product Certifications

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.