Learn More
Thermo Scientific™ Nalgene™ Color-Coded Unitary™ LDPE Wash Bottles
Use promo code "23291" to get your promotional price.
Description
High quality squeeze bottles with assorted color coded closures in the Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottle design. Bottle and dispensing straw are molded in one piece to prevent contamination when refilling. Wide mouth allows faster, easier filling. Color coded closures help with chemical identification and organization. Precision dispensing tip provides an ultra-fine, controlled stream without sputter; the tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic material for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.
- Polypropylene screw closure
- One bottle of each color closure (red, white, yellow and blue) included per package
- Bottle and tubulation are molded in one piece for uniform service
- Tip delivers a straight, uniform, ultrafine stream, but can also be cut back to increase flow
- Contents can be dispensed without tipping or shaking
- Bottle and Carton are recyclable in many communities
- Manufactured from non-toxic materials
- Flexible LDPE bottle provides soft, ergonomic squeeze
- Polypropylene screw closure
Includes:
A sample of Polypaper™ Right-to-Know Custom Labeling System
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Translucent |
Description | Color-Coded Unitary Wash Bottle Assortment |
Material | LDPE |
No. per Case | 16 |
No. per Pack | 4 |
Closure Color | Assorted (Red, White, Yellow, Blue) |
Closure Included | Yes |
Show More |