Learn More
Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems
Obtain a comprehensive solution when plastic is preferred to fiberboard.
Brand: Thermo Scientific™ CS509X24
200.09 EUR valid until 2024-10-31
Use promo code "21744" to get your promotional price.
This item is not returnable.
View return policy
Description
Obtain a comprehensive solution when plastic is preferred to fiberboard. Thermo Scientific™ Nalgene™ Cryogenic Storage Boxes for Locator™ and Locator Plus Systems fit in Thermo Scientific stainless-steel, ultra-low temperature freezer racks.
- Warranty varies by country, please contact us for details
Specifications
2 in. height: 1.0 and 1.5mL, 100 vials per box, case of 10 | |
1.5 mL | |
Vials (100) |
5.29 cu. ft. | |
Locator Jr. Plus, Locator 4 Plus, Locator 6 Plus |